Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000410.3 | NP_000401.1 | 5 | 1077 | ||
NM_001300749.1 | NP_001287678.1 | 5 | 1077 | |||
NM_139003.2 | NP_620572.1 | 4 | 759 | |||
NM_139004.2 | NP_620573.1 | 4 | 801 | |||
NM_139006.2 | NP_620575.1 | 5 | 1035 | |||
NM_139007.2 | NP_620576.1 | 4 | 813 | |||
NM_139008.2 | NP_620577.1 | 4 | 771 | |||
NM_139009.2 | NP_620578.1 | 5 | 1008 | |||
NM_139010.2 | NP_620579.1 | 3 | 537 | |||
NM_139011.2 | NP_620580.1 | 2 | 261 | |||
XM_011514543.2 | XP_011512845.1 | 5 | 1039 | |||
GenBank mRNA | AF079407.1 | - | 5 | 911 | ||
AF079408.1 | - | 4 | 689 | |||
AF079409.1 | - | 4 | 647 | |||
AF109385.1 | - | 1 | 327 | |||
AF115265.1 | - | 5 | 928 | |||
AJ249335.1 | - | 5 | 1008 | |||
AJ249336.1 | - | 4 | 813 | |||
AJ249337.1 | - | 4 | 801 | |||
AJ249338.1 | - | 2 | 261 | |||
AJ250635.1 | - | 3 | 537 | |||
AK300933.1 | - | 5 | 921 | |||
AY205604.1 | - | 4 | 678 | |||
BC074721.2 | - | 6 | 957 | 30915171 | ||
BC117201.1 | - | 5 | 952 | 40125752 | ||
BC117203.1 | - | 5 | 952 | 40125754 | ||
JN375755.1 | - | 2 | 135 | |||
JN375756.1 | - | 2 | 134 | |||
JN375757.1 | - | 2 | 125 | |||
JN375758.1 | - | 2 | 134 | |||
JN375759.1 | - | 2 | 140 | |||
U60319.1 | - | 5 | 1138 |