Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015594.2 | NP_056409.1 | 5 | 423 | ||
XM_011524596.1 | XP_011522898.1 | 5 | 349 | |||
GenBank mRNA | AL117394.1 | - | 6 | 2772 | ||
BC096718.1 | - | 5 | 423 | 40003088 | ||
BC098126.1 | - | 4 | 374 | 40003085 |