Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144564.4 | NP_653165.2 | 2 | 340 | ||
NM_213568.1 | NP_998733.1 | 2 | 340 | |||
GenBank mRNA | AK002044.1 | - | 2 | 303 | ||
AK302600.1 | - | 2 | 281 | |||
BC005869.2 | - | 2 | 272 | 2966557 | ||
BC020571.2 | - | 2 | 308 | |||
BG719479.1 | - | 2 | 317 | |||
BQ898834.1 | - | 416 |