Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPANXB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032461.3 | NP_115850.2 | 1 | 118 | ||
GenBank mRNA | AF098307.1 | - | 1 | 118 | ||
AJ457796.1 | - | 1 | 118 | |||
BC034472.1 | - | 1 | 116 | 4824918 | ||
BC137533.1 | - | 1 | 108 | 9021533 | ||
BC137536.1 | - | 1 | 108 | 9021536 |