Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025265.2 | NP_001020436.1 | 5, 6 | 665 | ||
NR_033212.1 | 5, 6 | 646 | ||||
XM_005262244.3 | XP_005262301.1 | 4, 5 | 781 | |||
GenBank mRNA | AA889552.1 | - | 1, 2 | 119 | ||
BC144434.1 | - | 5, 6 | 463 | |||
DB523273.1 | - | 4, 5 | 349 |