Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029859.2 | NP_001025030.1 | 2 | 246 | ||
XM_005273925.3 | XP_005273982.1 | 2 | 471 | |||
XM_005273928.1 | XP_005273985.1 | 3 | 371 | |||
XM_006718516.3 | XP_006718579.1 | 2 | 3416 | |||
XM_006718517.2 | XP_006718580.1 | 3 | 483 | |||
XM_006718518.3 | XP_006718581.1 | 2 | 231 | |||
XM_011544955.2 | XP_011543257.1 | 2 | 1711 | |||
GenBank mRNA | AK095233.1 | - | 2 | 243 | ||
AK300320.1 | - | 2 | 233 | |||
BC036058.1 | - | 2 | 235 | 5277643 | ||
DC342183.1 | - | 2 | 246 |
Gene Symbol: KCTD21-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC062331.1 | - | 1 | 617 | 5798762 |