Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004522.2 | NP_004513.1 | 1 | 24 | ||
XM_017004062.1 | XP_016859551.1 | 1 | 15 | |||
GenBank mRNA | AW162840.1 | - | 1 | 24 |