Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164586.1 | NP_001158058.1 | 12 | 1706 | ||
XM_005245580.3 | XP_005245637.1 | 12 | 1751 | |||
XM_006711618.3 | XP_006711681.1 | 12 | 1751 | |||
XM_011510120.2 | XP_011508422.1 | 12 | 1798 | |||
XM_011510121.2 | XP_011508423.1 | 12 | 1783 | |||
XM_017002787.1 | XP_016858276.1 | 11 | 1478 |