Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040424.2 | NP_001035514.1 | 24 | 6334 | ||
NM_001282934.1 | NP_001269863.1 | 25 | 6394 | |||
NR_104257.1 | 25 | 6342 | ||||
NR_104258.1 | 24 | 6222 | ||||
XM_006724039.2 | XP_006724102.1 | 24 | 6176 | |||
XM_006724040.2 | XP_006724103.1 | 23 | 6116 | |||
XM_011529674.1 | XP_011527976.1 | 24 | 6206 | |||
XM_011529675.1 | XP_011527977.1 | 25 | 6250 | |||
XM_011529676.2 | XP_011527978.1 | 25 | 8659 | |||
XM_011529677.2 | XP_011527979.1 | 25 | 6335 | |||
XM_011529678.2 | XP_011527980.1 | 25 | 6379 | |||
XM_011529679.2 | XP_011527981.1 | 25 | 6712 | |||
XM_011529681.2 | XP_011527983.1 | 25 | 7616 | |||
XM_011529683.1 | XP_011527985.1 | 30 | 7037 | |||
XM_017028425.1 | XP_016883914.1 | 24 | 6189 | |||
XM_017028426.1 | XP_016883915.1 | 24 | 8598 | |||
GenBank mRNA | AB051812.1 | - | 24 | 6325 | ||
AB051813.1 | - | 25 | 6385 | |||
AB051814.1 | - | 25 | 6333 | |||
AB051815.1 | - | 24 | 6213 | |||
AK123273.1 | - | 1 | 1682 | |||
AL355710.1 | - | 1 | 847 |