Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040424.2 | NP_001035514.1 | 24 | 6183 | ||
NM_001282934.1 | NP_001269863.1 | 25 | 6243 | |||
NR_104257.1 | 25 | 6191 | ||||
NR_104258.1 | 24 | 6071 | ||||
NR_104260.1 | 25 | 4598 | ||||
NR_135464.1 | 25 | 4739 | ||||
XM_006724039.2 | XP_006724102.1 | 24 | 6025 | |||
XM_006724040.2 | XP_006724103.1 | 23 | 5965 | |||
XM_011529674.1 | XP_011527976.1 | 24 | 6055 | |||
XM_011529675.1 | XP_011527977.1 | 25 | 6099 | |||
XM_011529676.2 | XP_011527978.1 | 25 | 8508 | |||
XM_011529677.2 | XP_011527979.1 | 25 | 6184 | |||
XM_011529678.2 | XP_011527980.1 | 25 | 6228 | |||
XM_011529679.2 | XP_011527981.1 | 25 | 6561 | |||
XM_011529681.2 | XP_011527983.1 | 25 | 7465 | |||
XM_011529683.1 | XP_011527985.1 | 30 | 6886 | |||
XM_017028425.1 | XP_016883914.1 | 24 | 6038 | |||
XM_017028426.1 | XP_016883915.1 | 24 | 8447 | |||
GenBank mRNA | AB051812.1 | - | 24 | 6174 | ||
AB051813.1 | - | 25 | 6234 | |||
AB051814.1 | - | 25 | 6182 | |||
AB051815.1 | - | 24 | 6062 | |||
AF426259.1 | - | 5 | 2191 | |||
AF426260.1 | - | 6 | 1898 | |||
AK123273.1 | - | 1 | 1531 | |||
AL355710.1 | - | 1 | 696 | |||
AY063456.1 | - | 7 | 2214 | |||
BC067102.1 | - | 25 | 4598 | 5925140 |