Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195280.1 | NP_001182209.1 | 2 | 216 | ||
XM_011515057.1 | XP_011513359.1 | 2 | 269 | |||
GenBank mRNA | AW341438.1 | - | 3 | 235 | ||
BF222754.1 | - | 2 | 237 | |||
DT932641.1 | - | 2 | 70 |