Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAG3L5P-PVRIG2P-PILRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036569.1 | 2 | 305 | |||
NR_036570.1 | 2 | 305 | ||||
GenBank mRNA | AK056412.1 | - | 2 | 256 | ||
AK310352.1 | - | 2 | 109 | |||
AL834317.1 | - | 2 | 127 | |||
DB275512.1 | - | 2 | 305 |
Gene Symbol: STAG3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040583.2 | 3 | 381 | |||
GenBank mRNA | AK289466.1 | - | 4 | 350 | ||
AK291820.1 | - | 1 | 21 | |||
BG829469.1 | - | 2 | 265 | |||
DC419304.1 | - | 2 | 359 |
Gene Symbol: STAG3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040584.2 | 2 | 257 | |||
GenBank mRNA | AK303473.1 | - | 3 | 211 | ||
AK307228.1 | - | 3 | 328 | |||
AK309007.1 | - | 3 | 212 | |||
BC002581.2 | - | 3 | 199 | |||
BG537494.1 | - | 2 | 256 | |||
BI753182.1 | - | 3 | 202 | |||
CB956482.1 | - | 2 | 236 |
Gene Symbol: STAG3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040582.2 | 2 | 400 | |||
GenBank mRNA | AK303592.1 | - | 2 | 61 | ||
AK316130.1 | - | 2 | 263 | |||
AY313779.1 | - | 2 | 255 | |||
BC066937.1 | - | 2 | 133 | 4826886 | ||
BC108680.1 | - | 2 | 267 | |||
BI829811.1 | - | 2 | 135 | |||
BM007884.1 | - | 2 | 194 | |||
DA505391.1 | - | 2 | 272 | |||
DA555040.1 | - | 2 | 282 | |||
DC395490.1 | - | 2 | 183 | |||
DC405605.1 | - | 2 | 382 | |||
DC410426.1 | - | 2 | 61 |
Gene Symbol: STAG3L5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103720.1 | 2 | 174 | |||
GenBank mRNA | BM826568.1 | - | 2 | 128 | ||
DA306290.1 | - | 2 | 174 |
Gene Symbol: TRIM74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012197.1 | XP_016867686.1 | 2 | 272 |