Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014249.3 | NP_055064.1 | 2 | 368 | ||
NM_016346.3 | NP_057430.1 | 2 | 368 | |||
GenBank mRNA | AB307710.1 | - | 2 | 172 | ||
AF121129.1 | - | 2 | 362 | |||
AF148128.1 | - | 2 | 325 | |||
AL711747.1 | - | 2 | 369 | |||
BC041421.1 | - | 4 | 398 | 3876974 | ||
HQ692847.1 | - | 2 | 172 |