Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308176.1 | NP_001295105.1 | 4 | 564 | ||
NM_001792.4 | NP_001783.2 | 5 | 1044 | |||
XM_011525788.1 | XP_011524090.1 | 5 | 709 | |||
XM_017025514.1 | XP_016881003.1 | 5 | 1015 | |||
GenBank mRNA | AK302831.1 | - | 4 | 554 | ||
BC036470.1 | - | 5 | 736 | 5264944 | ||
S42303.1 | - | 5 | 1078 | |||
X54315.1 | - | 5 | 721 |