Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139444.2 | NP_001132916.1 | 5 | 542 | ||
GenBank mRNA | AI745276.1 | - | 1 | 462 | ||
AK002042.1 | - | 3 | 356 | |||
BC127757.1 | - | 2 | 273 | 40127548 | ||
BC127758.1 | - | 2 | 272 | 40127555 | ||
DY655209.1 | - | 4 | 349 |