Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SV2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167580.1 | NP_001161052.1 | 12 | 6296 | ||
NM_001323031.1 | NP_001309960.1 | 13 | 6829 | |||
NM_001323032.1 | NP_001309961.1 | 13 | 7138 | |||
NM_001323033.1 | NP_001309962.1 | 11 | 6805 | |||
NM_001323034.1 | NP_001309963.1 | 12 | 6937 | |||
NM_001323036.1 | NP_001309965.1 | 12 | 6115 | |||
NM_001323037.1 | NP_001309966.1 | 14 | 7045 | |||
NM_001323038.1 | NP_001309967.1 | 13 | 6957 | |||
NM_001323039.1 | NP_001309968.1 | 13 | 7230 | |||
NM_001323040.1 | NP_001309969.1 | 13 | 6476 | |||
NM_014848.5 | NP_055663.1 | 14 | 7226 | |||
XM_005254998.3 | XP_005255055.1 | 13 | 6891 | |||
XM_017022761.1 | XP_016878250.1 | 13 | 6842 | |||
XM_017022762.1 | XP_016878251.1 | 13 | 6977 | |||
GenBank mRNA | AK124722.1 | - | 1 | 887 |