Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005247167.1 | XP_005247224.1 | 1 | 219 | ||
XM_011512490.2 | XP_011510792.1 | 1 | 219 | |||
XM_011512491.2 | XP_011510793.1 | 1 | 219 | |||
XM_011512492.2 | XP_011510794.1 | 1 | 219 |