Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101060179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003959932.4 | XP_003959981.1 | 1 | 309 |
Gene Symbol: PRDM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011520230.2 | XP_011518532.1 | 7 | 2261 | ||
XM_017018024.1 | XP_016873513.1 | 5 | 1564 | |||
GenBank mRNA | AK097878.1 | - | 2 | 505 | ||
AY726563.1 | - | 2 | 1212 |