Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011526076.2 | XP_011524378.1 | 1 | 437 | ||
GenBank mRNA | BX648603.1 | - | 1 | 61 |