Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278700.1 | NP_001265629.1 | 1 | 376 | ||
XM_006713666.2 | XP_006713729.1 | 1 | 371 | |||
XM_006713667.2 | XP_006713730.1 | 1 | 363 | |||
XM_006713668.3 | XP_006713731.1 | 1 | 380 | |||
XM_006713670.2 | XP_006713733.1 | 1 | 371 | |||
XM_017006636.1 | XP_016862125.1 | 1 | 371 | |||
XM_017006637.1 | XP_016862126.1 | 1 | 371 | |||
GenBank mRNA | BM541682.1 | - | 1 | 333 | ||
HY178178.1 | - | 1 | 376 |