Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278698.1 | NP_001265627.1 | 1 | 117 | ||
NM_001278699.1 | NP_001265628.1 | 1 | 117 | |||
NM_001278714.1 | NP_001265643.1 | 1 | 117 | |||
NM_001278715.1 | NP_001265644.1 | 1 | 117 | |||
NM_006232.3 | NP_006223.2 | 1 | 117 | |||
XM_006713666.2 | XP_006713729.1 | 2 | 655 | |||
XM_006713668.3 | XP_006713731.1 | 2 | 664 | |||
XM_006713670.2 | XP_006713733.1 | 2 | 655 | |||
XM_017006636.1 | XP_016862125.1 | 2 | 655 | |||
XM_017006637.1 | XP_016862126.1 | 2 | 655 | |||
GenBank mRNA | CN368789.1 | - | 1 | 121 |