Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005256038.3 | XP_005256095.1 | 19 | 5199 | ||
GenBank mRNA | AK090627.1 | - | 1 | 422 | ||
AK128439.1 | - | 1 | 1744 |