Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM65C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290268.1 | NP_001277197.1 | 1 | 282 | ||
NR_110890.1 | 1 | 282 | ||||
XM_005260294.3 | XP_005260351.1 | 2 | 279 | |||
XM_006723713.3 | XP_006723776.1 | 1 | 342 | |||
XM_011528578.2 | XP_011526880.1 | 2 | 307 | |||
XM_011528579.2 | XP_011526881.1 | 1 | 342 | |||
XM_011528580.2 | XP_011526882.1 | 1 | 342 | |||
XM_011528581.2 | XP_011526883.1 | 1 | 342 | |||
XM_011528584.2 | XP_011526886.1 | 1 | 344 | |||
XM_011528586.2 | XP_011526888.1 | 1 | 345 | |||
XM_017027682.1 | XP_016883171.1 | 1 | 344 | |||
GenBank mRNA | AK124347.1 | - | 1 | 294 | ||
AK299337.1 | - | 1 | 131 | |||
BM846795.1 | - | 1 | 278 | |||
DC400584.1 | - | 1 | 283 |