Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGAP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027032.1 | 2 | 485 | |||
GenBank mRNA | AK301303.1 | - | 2 | 459 | ||
BC039697.1 | - | 2 | 404 | |||
CN313469.1 | - | 2 | 485 |