Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006722580.3 | XP_006722643.1 | 2 | 369 | ||
XM_011526263.2 | XP_011524565.1 | 3 | 461 | |||
XM_017026093.1 | XP_016881582.1 | 3 | 461 | |||
GenBank mRNA | AK094926.1 | - | 2 | 373 |