Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAC14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034083.2 | 2 | 245 | |||
GenBank mRNA | AK095020.1 | - | 2 | 171 | ||
BC035152.1 | - | 2 | 210 | |||
DB487352.1 | - | 2 | 245 |