Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300LD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115152.1 | NP_001108624.1 | 2 | 128 | ||
GenBank mRNA | AY358148.1 | - | 2 | 184 | ||
EF137868.1 | - | 2 | 128 |