Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010881.1 | NP_001010881.1 | 3 | 289 | ||
XM_011541269.2 | XP_011539571.2 | 4 | 536 | |||
XM_011541271.2 | XP_011539573.2 | 4 | 385 | |||
XM_011541272.2 | XP_011539574.1 | 4 | 385 | |||
XM_011541274.2 | XP_011539576.2 | 3 | 403 | |||
XM_011541275.2 | XP_011539577.2 | 3 | 200 | |||
XM_011541276.2 | XP_011539578.1 | 4 | 385 | |||
XM_011541277.2 | XP_011539579.1 | 4 | 385 | |||
XM_011541278.2 | XP_011539580.1 | 4 | 385 | |||
XM_017001062.1 | XP_016856551.1 | 5 | 523 | |||
XM_017001063.1 | XP_016856552.1 | 4 | 376 | |||
XM_017001064.1 | XP_016856553.1 | 4 | 385 | |||
GenBank mRNA | BC128091.1 | - | 2 | 102 | ||
BC128092.1 | - | 2 | 102 | |||
HY056181.1 | - | 3 | 255 |