Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114107.4 | NP_001107579.1 | 7 | 2245 | ||
NM_001257962.1 | NP_001244891.1 | 7 | 2125 | |||
NM_001257963.1 | NP_001244892.1 | 5 | 1888 | |||
NM_014476.5 | NP_055291.2 | 8 | 2389 | |||
NR_047562.1 | 6 | 2057 | ||||
GenBank mRNA | AK129607.1 | - | 1 | 378 |