Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PET100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171155.1 | NP_001164626.1 | 2 | 98 | ||
NR_033242.1 | 2 | 98 | ||||
GenBank mRNA | AK124717.1 | - | 1 | 786 | ||
AW009489.1 | - | 3 | 234 | |||
BC150496.1 | - | 2 | 76 | |||
BU740873.1 | - | 3 | 256 | |||
CB119056.1 | - | 2 | 89 | |||
CD641681.1 | - | 2 | 92 |