Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104599.1 | 3 | 760 | |||
GenBank mRNA | AK000440.1 | - | 3 | 660 | ||
BC039252.1 | - | 3 | 660 | |||
BC065002.1 | - | 3 | 630 |