Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121630.1 | 2 | 384 | |||
NR_121631.1 | 2 | 156 | ||||
NR_121632.1 | 2 | 173 | ||||
NR_121633.1 | 2 | 173 | ||||
GenBank mRNA | AW172333.1 | - | 2 | 266 | ||
AW611902.1 | - | 1 | 39 | |||
BM713112.1 | - | 1 | 64 | |||
BM717037.1 | - | 1 | 64 | |||
DA940436.1 | - | 2 | 156 | |||
DB177224.1 | - | 2 | 243 |