Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSC-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033651.1 | 2 | 345 | |||
NR_033652.1 | 3 | 814 | ||||
GenBank mRNA | AK123315.1 | - | 3 | 174 | ||
AK304427.1 | - | 3 | 814 | |||
BC094782.1 | - | 2 | 385 | |||
BX647470.1 | - | 2 | 358 | |||
DB161440.1 | - | 2 | 315 |