Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM159B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164442.1 | NP_001157914.1 | 1, 2 | 509 | ||
GenBank mRNA | BQ271827.1 | - | 1, 2 | 509 | ||
BU785751.1 | - | 1, 2 | 289 | |||
BU952508.1 | - | 2, 3 | 457 |