Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097872.1 | - | 2 | 446 | ||
AK296003.1 | - | 6 | 1004 | |||
AK298260.1 | - | 6 | 1090 |