Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130918.1 | NP_001124390.1 | 2 | 199 | ||
XM_017024491.1 | XP_016879980.1 | 2 | 345 | |||
XM_017024493.1 | XP_016879982.1 | 3 | 406 | |||
XM_017024494.1 | XP_016879983.1 | 2 | 341 | |||
XM_017024495.1 | XP_016879984.1 | 1 | 22 | |||
XM_017024497.1 | XP_016879986.1 | 2 | 169 | |||
GenBank mRNA | AK097392.1 | - | 2 | 111 | ||
AK302952.1 | - | 2 | 145 | |||
AL834151.1 | - | 2 | 170 | |||
BC041368.2 | - | 2 | 145 |