Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017024492.1 | XP_016879981.1 | 2 | 232 | ||
XM_017024495.1 | XP_016879984.1 | 1 | 642 | |||
XM_017024497.1 | XP_016879986.1 | 3 | 541 | |||
GenBank mRNA | AL834151.1 | - | 3 | 542 | ||
BC041368.2 | - | 2 | 765 |