Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317812.1 | NP_001304741.1 | 4 | 3646 | ||
NM_001317813.1 | NP_001304742.1 | 5 | 3293 | |||
NM_001317814.1 | NP_001304743.1 | 5 | 3240 | |||
NM_016612.3 | NP_057696.2 | 4 | 3169 | |||
GenBank mRNA | AF495725.1 | - | 1 | 1156 | ||
BX647289.1 | - | 1 | 282 |