Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYO15B SIRNA Assays ›
Gene Symbol:
MYO15B
Gene Name:
myosin XVB
Gene Aliases:
MYO15BP
Chromosome Location:
Chr.17: 75587545 - 75626850 on Build GRCh38
UniGene ID:
Hs.390817
Species:
Human
siRNA ID
HSS173134
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYO15B
Interrogated Sequence
Translated Protein
RNADB ID
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001309242.1
NP_001296171.1
59, 60
8694
XM_017025120.1
XP_016880609.1
61, 62
8401
XM_017025121.1
XP_016880610.1
61, 62
8386
XM_017025122.1
XP_016880611.1
61, 62
8365
XM_017025123.1
XP_016880612.1
61, 62
8337
XM_017025124.1
XP_016880613.1
60, 61
8275
XM_017025125.1
XP_016880614.1
59, 60
8124
XM_017025126.1
XP_016880615.1
56, 57
7866
XM_017025127.1
XP_016880616.1
59, 60
7709
XM_017025128.1
XP_016880617.1
60, 61
7455
XM_017025129.1
XP_016880618.1
59, 60
6687
XM_017025130.1
XP_016880619.1
58, 59
6643
XM_017025131.1
XP_016880620.1
57, 58
7082
XM_017025132.1
XP_016880621.1
56, 57
7023
XM_017025133.1
XP_016880622.1
54, 55
6548
XM_017025134.1
XP_016880623.1
55, 56
6909
XM_017025135.1
XP_016880624.1
49, 50
5565
XM_017025136.1
XP_016880625.1
49, 50
5914
XM_017025137.1
XP_016880626.1
46, 47
5459
XM_017025138.1
XP_016880627.1
41, 42
4955
XM_017025139.1
XP_016880628.1
42, 43
4756
XM_017025140.1
XP_016880629.1
39, 40
4569
XM_017025143.1
XP_016880632.1
34, 35
3846
XM_017025144.1
XP_016880633.1
55, 56
8329
GenBank mRNA
AF418292.1
-
34, 35
4304
AF418293.1
-
35, 36
3910
AK026339.1
-
11, 12
1009
AK096869.1
-
11, 12
1148
AL713730.1
-
10, 11
903
BC027875.1
-
LIT3316
MYO15BP
11, 12
1398
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline