Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286555.1 | NP_001273484.1 | 7 | 1087 | ||
NR_104473.1 | 6 | 1037 | ||||
NR_104474.1 | 6 | 1004 | ||||
NR_104475.1 | 4 | 920 | ||||
XM_011514757.2 | XP_011513059.1 | 4 | 549 | |||
GenBank mRNA | AB208997.1 | - | 4 | 440 | ||
AK000383.1 | - | 3 | 413 | |||
AK074410.1 | - | 3 | 413 | |||
AK128760.1 | - | 4 | 1408 | |||
AK296402.1 | - | 7 | 657 | |||
AK310653.1 | - | 5 | 1110 | |||
BX362191.2 | - | 4 | 701 | |||
BX648953.1 | - | 4 | 5086 | |||
DB177174.1 | - | 6 | 541 |