Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284303.1 | NP_001271232.1 | 11 | 2354 | ||
NM_001284304.1 | NP_001271233.1 | 14 | 2725 | |||
NM_001284305.1 | NP_001271234.1 | 13 | 2665 | |||
NM_014346.3 | NP_055161.1 | 13 | 2588 | |||
NR_104292.1 | 14 | 3034 | ||||
XM_017028742.1 | XP_016884231.1 | 14 | 2680 | |||
XM_017028744.1 | XP_016884233.1 | 14 | 2576 | |||
XM_017028745.1 | XP_016884234.1 | 12 | 2407 | |||
GenBank mRNA | AK000851.1 | - | 4 | 1259 |