Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015053.1 | NP_001015053.1 | 23 | 3186 | ||
NM_005474.4 | NP_005465.2 | 23 | 3183 | |||
XM_005256905.1 | XP_005256962.1 | 23 | 3181 | |||
XM_005256906.4 | XP_005256963.1 | 23 | 3142 | |||
XM_011524149.1 | XP_011522451.1 | 23 | 3157 | |||
XM_011524150.1 | XP_011522452.1 | 22 | 2933 | |||
XM_017023988.1 | XP_016879477.1 | 25 | 3295 | |||
XM_017023989.1 | XP_016879478.1 | 25 | 3261 | |||
XM_017023990.1 | XP_016879479.1 | 25 | 3286 | |||
XM_017023991.1 | XP_016879480.1 | 25 | 3292 | |||
XM_017023992.1 | XP_016879481.1 | 25 | 3281 | |||
XM_017023993.1 | XP_016879482.1 | 25 | 3268 | |||
XM_017023994.1 | XP_016879483.1 | 21 | 3249 | |||
XM_017023995.1 | XP_016879484.1 | 23 | 3151 | |||
XM_017023996.1 | XP_016879485.1 | 24 | 3358 | |||
GenBank mRNA | AB011172.2 | - | 21 | 2900 | ||
AB177853.1 | - | 13 | 1635 | |||
AF039691.1 | - | 18 | 2288 | |||
AF132608.1 | - | 22 | 2851 | |||
BC038423.1 | - | 12 | 1465 | |||
BC051824.1 | - | 23 | 3062 | 6043491 |