Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L11 SIRNA Assays ›
Gene Symbol:
BCL2L11
Gene Name:
BCL2 like 11
Gene Aliases:
BAM, BIM, BOD
Chromosome Location:
Chr.2: 111120914 - 111168445 on Build GRCh38
UniGene ID:
Hs.469658
Species:
Human
siRNA ID
HSS173329
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204106.1
NP_001191035.1
4
564
NM_001204107.1
NP_001191036.1
6
761
NM_001204108.1
NP_001191037.1
5
966
NM_001204109.1
NP_001191038.1
4
855
NM_001204110.1
NP_001191039.1
5
696
NM_001204111.1
NP_001191040.1
4
550
NM_001204112.1
NP_001191041.1
5
675
NM_006538.4
NP_006529.1
5
654
NM_138621.4
NP_619527.1
4
834
NM_138622.3
NP_619528.1
5
959
NM_138623.3
NP_619529.1
6
779
NM_138624.3
NP_619530.1
5
867
NM_138625.3
NP_619531.1
5
680
NM_138626.3
NP_619532.1
3
730
NM_138627.3
NP_619533.1
3
460
NM_207003.2
NP_996886.1
5
689
XM_005263551.2
XP_005263608.1
4
1612
XM_005263553.2
XP_005263610.1
5
1432
XM_005263554.2
XP_005263611.1
3
1508
XM_005263559.2
XP_005263616.1
4
1342
XM_017003101.1
XP_016858590.1
4
1328
GenBank mRNA
AB071195.1
-
4
671
AB071196.1
-
5
491
AB071197.1
-
4
579
AB071198.1
-
2
442
AB071199.1
-
4
392
AB071200.1
-
2
172
AF032457.1
-
3
546
AF032458.1
-
4
366
AK290377.1
-
5
654
AK291269.1
-
4
831
AY305714.1
-
3
276
AY305715.1
-
4
401
AY305716.1
-
4
298
AY423441.1
-
4
401
AY423442.1
-
5
473
AY423443.1
-
4
678
AY428962.1
-
3
567
BC033694.1
-
4
816
5213713
DQ849200.1
-
4
408
DQ849201.1
-
3
262
DQ849202.1
-
4
387
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
B cell homeostasis
B cell apoptotic process
kidney development
protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
myeloid cell homeostasis
apoptotic process
meiosis I
cell-matrix adhesion
spermatogenesis
brain development
male gonad development
intrinsic apoptotic signaling pathway in response to DNA damage
mammary gland development
positive regulation of protein homooligomerization
response to endoplasmic reticulum stress
tube formation
odontogenesis of dentin-containing tooth
T cell homeostasis
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of neuron apoptotic process
ear development
positive regulation of cell cycle
regulation of organ growth
developmental pigmentation
regulation of developmental pigmentation
spleen development
thymus development
post-embryonic organ morphogenesis
positive regulation of apoptotic process by virus
cellular process regulating host cell cycle in response to virus
thymocyte apoptotic process
positive regulation of release of cytochrome c from mitochondria
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
apoptotic process involved in embryonic digit morphogenesis
positive regulation of IRE1-mediated unfolded protein response
positive regulation of fibroblast apoptotic process
positive regulation of intrinsic apoptotic signaling pathway
protein binding
microtubule binding
dynein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-558c998fd6-nvdhv:80/10.239.163.173:80.
git-commit: a0437b286f8214373ac979cfd76c0df1857d95b8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.14.2-2024.08.97-1.0