Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TROAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100620.1 | NP_001094090.1 | 3 | 393 | ||
NM_001278324.1 | NP_001265253.1 | 3 | 393 | |||
NM_005480.3 | NP_005471.3 | 3 | 393 | |||
XM_006719181.1 | XP_006719244.1 | 3 | 337 | |||
XM_011537723.1 | XP_011536025.1 | 3 | 337 | |||
XM_011537725.2 | XP_011536027.1 | 4 | 529 | |||
GenBank mRNA | AK296811.1 | - | 3 | 326 | ||
AK299302.1 | - | 3 | 298 | |||
AK315211.1 | - | 3 | 337 | |||
BC011597.2 | - | 3 | 261 | |||
BC032583.1 | - | 3 | 305 | 5498800 | ||
CD366215.1 | - | 1 | 475 | |||
DC410015.1 | - | 3 | 393 | |||
U04810.2 | - | 3 | 336 |