Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005482.2 | NP_005473.1 | 10 | 1050 | ||
GenBank mRNA | AF022913.1 | - | 10 | 1022 | ||
AK304340.1 | - | 8 | 805 | |||
AK313015.1 | - | 10 | 1033 | |||
BC020737.1 | - | 10 | 1029 | 4722399 | ||
BX648473.1 | - | 7 | 752 | |||
Y07596.1 | - | 10 | 1009 |