Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005689.2 | NP_005680.1 | 14 | 2248 | ||
GenBank mRNA | AF070598.1 | - | 10 | 940 | ||
AF076775.1 | - | 14 | 1931 | |||
AF308472.1 | - | 14 | 2237 | |||
AJ289233.2 | - | 14 | 2208 | |||
AK026067.1 | - | 11 | 1057 | |||
AK057026.1 | - | 14 | 1967 | |||
AK172812.1 | - | 14 | 1598 | |||
BC000559.2 | - | 14 | 2199 | 3162881 | ||
BC043423.1 | - | 7 | 2248 | |||
U66673.1 | - | 3 | 224 |