Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256670.1 | NP_001243599.1 | 9 | 1383 | ||
NM_005700.4 | NP_005691.2 | 10 | 1473 | |||
NM_130443.3 | NP_569710.2 | 10 | 1476 | |||
GenBank mRNA | AB017970.1 | - | 4 | 286 | ||
AJ271216.1 | - | 10 | 1072 | |||
AK025984.1 | - | 6 | 481 | |||
AK222566.1 | - | 10 | 1050 | |||
AK222847.1 | - | 10 | 1051 | |||
AK297199.1 | - | 9 | 958 | |||
AK304581.1 | - | 11 | 974 | |||
AK315478.1 | - | 10 | 1464 | |||
AL833475.1 | - | 10 | 1078 | |||
BC001446.2 | - | 9 | 1036 | 3138626 | ||
BC007221.1 | - | 10 | 1062 | 3349135 | ||
BC014038.2 | - | 10 | 1036 | 4547232 | ||
BC024271.1 | - | 9 | 1032 | 4564651 |