Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005707.1 | NP_005698.1 | 2 | 1010 | ||
GenBank mRNA | AF083930.1 | - | 2 | 155 | ||
AK096970.1 | - | 2 | 1010 | |||
AK225816.1 | - | 2 | 977 | |||
BC092464.1 | - | 4 | 334 | |||
BC131704.1 | - | 3 | 312 |