Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005707.1 | NP_005698.1 | 1, 2 | 925 | ||
GenBank mRNA | AK096970.1 | - | 1, 2 | 925 | ||
AK225816.1 | - | 1, 2 | 892 | |||
BC092464.1 | - | 3, 4 | 249 | |||
BC131704.1 | - | 2, 3 | 227 |