Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270439.1 | NP_001257368.1 | 4 | 711 | ||
NM_005717.3 | NP_005708.1 | 4 | 702 | |||
GenBank mRNA | AF006088.1 | - | 4 | 444 | ||
AF017807.1 | - | 4 | 423 | |||
AK095424.1 | - | 3 | 1082 | |||
AK129598.1 | - | 4 | 487 | |||
BC057237.1 | - | 4 | 531 | 3931697 | ||
BI758106.1 | - | 4 | 612 | |||
CN291383.1 | - | 4 | 702 |